Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018398.2 | Intron | NP_060868.2 | |||
XM_005265318.2 | Intron | XP_005265375.1 | |||
XM_011533946.2 | Intron | XP_011532248.1 | |||
XM_011533947.2 | Intron | XP_011532249.1 | |||
XM_011533948.2 | Intron | XP_011532250.1 | |||
XM_011533949.2 | Intron | XP_011532251.1 | |||
XM_011533950.2 | Intron | XP_011532252.1 | |||
XM_011533951.2 | Intron | XP_011532253.1 | |||
XM_011533952.2 | Intron | XP_011532254.1 | |||
XM_011533953.2 | Intron | XP_011532255.1 | |||
XM_011533954.2 | Intron | XP_011532256.1 | |||
XM_011533955.1 | Intron | XP_011532257.1 | |||
XM_017006850.1 | Intron | XP_016862339.1 | |||
XM_017006851.1 | Intron | XP_016862340.1 | |||
XM_017006852.1 | Intron | XP_016862341.1 | |||
XM_017006853.1 | Intron | XP_016862342.1 | |||
XM_017006854.1 | Intron | XP_016862343.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001304389.1 | Intron | NP_001291318.1 | |||
NM_020678.3 | Intron | NP_065729.1 |