Product Details

SNP ID
rs522376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58938591 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGATATCAGAACTAAGCTAGGCCC[A/G]GAGAGCCCTGAGGTTGTGACACATA
Phenotype
MIM: 614761
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLYATL1 PubMed Links

Gene Details

Gene
GLYATL1
Gene Name
glycine-N-acyltransferase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220494.2 Intron NP_001207423.1
NM_001220496.2 Intron NP_001207425.1
NM_080661.4 Intron NP_542392.2
XM_011545354.2 Intron XP_011543656.1
XM_011545355.2 Intron XP_011543657.1
XM_011545356.2 Intron XP_011543658.1
XM_011545357.2 Intron XP_011543659.1
XM_011545358.2 Intron XP_011543660.1
XM_011545359.2 Intron XP_011543661.1
Gene
LOC283194
Gene Name
uncharacterized LOC283194
There are no transcripts associated with this gene.

View Full Product Details