Product Details

SNP ID
rs158103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77101322 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCAGTGCTGCTACTCCTCCGAAA[C/G]AGTAAAGTTTGGGGAGGGATGTAGC
Phenotype
MIM: 616995
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CIPC PubMed Links

Gene Details

Gene
CIPC
Gene Name
CLOCK interacting pacemaker
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033426.2 Intron NP_219494.2

View Full Product Details