Product Details

SNP ID
rs341790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:8982462 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTAGGTATTTGTTTTTACATTGAT[A/C]TATCTACTAGGTGCAAAGATTTGGT
Phenotype
MIM: 606525
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SRGAP3 PubMed Links

Gene Details

Gene
SRGAP3
Gene Name
SLIT-ROBO Rho GTPase activating protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033117.2 7106 UTR 3 NP_001028289.1
NM_014850.3 7106 UTR 3 NP_055665.1
XM_011534295.1 7106 UTR 3 XP_011532597.1
XM_011534296.1 7106 UTR 3 XP_011532598.1
XM_011534301.2 7106 UTR 3 XP_011532603.1
XM_011534303.1 7106 UTR 3 XP_011532605.1
XM_017007574.1 7106 UTR 3 XP_016863063.1
XM_017007575.1 7106 UTR 3 XP_016863064.1
XM_017007576.1 7106 UTR 3 XP_016863065.1
XM_017007577.1 7106 UTR 3 XP_016863066.1
XM_017007578.1 7106 UTR 3 XP_016863067.1
XM_017007579.1 7106 UTR 3 XP_016863068.1
XM_017007580.1 7106 Intron XP_016863069.1

View Full Product Details