Product Details

SNP ID
rs192752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:26137595 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTAGAAGGAATTATGTTGAAAGAA[A/T]TAATATATAAGAATATGTCTCACTA
Phenotype
MIM: 604242
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RNF6 PubMed Links

Gene Details

Gene
RNF6
Gene Name
ring finger protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005977.3 Intron NP_005968.1
NM_183043.2 Intron NP_898864.1
NM_183044.2 Intron NP_898865.1
NM_183045.1 Intron NP_898866.1
XM_005266485.2 Intron XP_005266542.1
XM_005266486.2 Intron XP_005266543.1
XM_011535177.2 Intron XP_011533479.1
XM_011535178.1 Intron XP_011533480.1
XM_017020685.1 Intron XP_016876174.1
XM_017020686.1 Intron XP_016876175.1
XM_017020687.1 Intron XP_016876176.1

View Full Product Details