Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nac-1
Entrez Gene ID
181585
Gene Name
Sodium-dependent low-affinity dicarboxylate transporter 1
Gene Aliases
CELE_F31F6.6
Chromosome Location
Chr.X: 14895145 - 14898497 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078072.5 NP_510473.2 10 - 11 1572 68
GenBank mRNA AY090484.1 - 10 - 11 1582 68

View Full Product Details