Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
taf-4
Entrez Gene ID
171630
Gene Name
TAFH domain-containing protein
Gene Aliases
CELE_R119.6
Chromosome Location
Chr.I: 377186 - 388574 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058327.8 NP_490728.2 6 - 7 1389 67

View Full Product Details