Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gbb-1
Entrez Gene ID
189837
Gene Name
protein-coding
Gene Aliases
CELE_Y41G9A.4
Chromosome Location
Chr.X: 2957934 - 2968194 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270068.6 NP_001256997.2 3 - 4 481 96
NM_001313456.4 NP_001300385.1 3 - 3 463 96
NM_171645.6 NP_741740.3 3 - 4 480 96
NM_171646.5 NP_741741.2 3 - 4 482 96
GenBank mRNA EU729334.1 - 3 - 4 463 96

View Full Product Details