Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-9
Entrez Gene ID
181443
Gene Name
protein-coding
Gene Aliases
CELE_R12H7.1
Chromosome Location
Chr.X: 13201770 - 13216729 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330917.4 NP_001317908.1 4 - 5 749 83
NM_171790.7 NP_741917.1 7 - 8 1067 83
GenBank mRNA U93195.1 - 7 - 8 1065 83

View Full Product Details