Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gar-2
Entrez Gene ID
175893
Gene Name
protein-coding
Gene Aliases
CELE_F47D12.1
Chromosome Location
Chr.III: 6304490 - 6320872 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027422.4 NP_001022593.1 11 - 12 1641 101
NM_001379757.2 NP_001368076.1 12 - 13 1888 212
GenBank mRNA AY053364.1 - 10 - 11 1709 212
AY053365.1 - 11 - 12 1598 101

View Full Product Details