Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lact-3
Entrez Gene ID
174644
Gene Name
Beta-lactamase domain-containing protein
Gene Aliases
CELE_M28.6
Chromosome Location
Chr.II: 10652353 - 10657841 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063898.4 NP_496299.1 5 - 6 1074 62

Gene Symbol
M28.5
Entrez Gene ID
174645
Gene Name
NHP2-like protein 1 homolog
Gene Aliases
CELE_M28.5
Chromosome Location
Chr.II: 10648892 - 10658855 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381581.2 NP_001366718.1 7 - 8 1874 62

View Full Product Details