Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-64
Entrez Gene ID
185264
Gene Name
C-type LECtin
Gene Aliases
CELE_F35C5.7
Chromosome Location
Chr.II: 12898242 - 12900439 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064343.3 NP_496744.1 2 - 3 706 114

View Full Product Details