Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
daf-5
Entrez Gene ID
175054
Gene Name
c-SKI_SMAD_bind domain-containing protein
Gene Aliases
CELE_W01G7.1
Chromosome Location
Chr.II: 14033216 - 14037721 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064540.6 NP_496941.1 1 - 2 255 109
GenBank mRNA AY438643.1 - 1 - 2 255 109

View Full Product Details