Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gei-4
Entrez Gene ID
175195
Gene Name
protein-coding
Gene Aliases
CELE_W07B3.2
Chromosome Location
Chr.III: 344306 - 418744 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001384004.1 21 - 22 3087 120
NM_001384005.1 3 - 4 360 120
NM_064789.5 NP_497190.1 7 - 8 1119 120

View Full Product Details