Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-29
Entrez Gene ID
184312
Gene Name
Prion-like-(Q/N-rich)-domain-bearing protein
Gene Aliases
CELE_F10F2.9
Chromosome Location
Chr.III: 4643929 - 4646346 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065547.6 NP_497948.1 3 - 4 391 74

View Full Product Details