Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wht-7
Entrez Gene ID
175864
Gene Name
protein-coding
Gene Aliases
CELE_Y42G9A.6
Chromosome Location
Chr.III: 6141959 - 6149597 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379747.1 NP_001367851.1 1 - 2 101 107

View Full Product Details