Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ani-1
Entrez Gene ID
176672
Gene Name
Anillin-like protein 1
Gene Aliases
CELE_Y49E10.19
Chromosome Location
Chr.III: 12446353 - 12453204 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067223.8 NP_499624.2 3 - 4 1121 86

View Full Product Details