Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-8
Entrez Gene ID
183207
Gene Name
Zinc metalloproteinase nas-8
Gene Aliases
CELE_C34D4.9
Chromosome Location
Chr.IV: 7114899 - 7119926 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068713.7 NP_501114.2 7 - 8 814 67

View Full Product Details