Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cwn-2
Entrez Gene ID
177870
Gene Name
Protein Wnt-2
Gene Aliases
CELE_W01B6.1
Chromosome Location
Chr.IV: 10067816 - 10070251 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069421.6 NP_501822.1 3 - 4 683 121

View Full Product Details