Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pfd-1
Entrez Gene ID
178021
Gene Name
protein-coding
Gene Aliases
CELE_C08F8.1
Chromosome Location
Chr.IV: 11135658 - 11150932 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268611.3 NP_001255540.1 2 - 3 338 103

View Full Product Details