Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vha-19
Entrez Gene ID
177103
Gene Name
Vacuolar H ATPase
Gene Aliases
CELE_Y55H10A.1
Chromosome Location
Chr.IV: 3369197 - 3373485 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067931.5 NP_500332.1 5 - 6 962 82

View Full Product Details