Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bec-1
Entrez Gene ID
177345
Gene Name
protein-coding
Gene Aliases
CELE_T19E7.3
Chromosome Location
Chr.IV: 5661141 - 5664425 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380239.1 NP_001368359.1 1 - 2 171 98
NM_068443.4 NP_500844.1 6 - 7 950 98

View Full Product Details