Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scl-10
Entrez Gene ID
186049
Gene Name
SCP domain-containing protein
Gene Aliases
CELE_F49E11.5
Chromosome Location
Chr.IV: 13048457 - 13049859 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070097.2 NP_502498.1 2 - 3 484 130

View Full Product Details