The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_070951.5 | NP_503352.1 | 5 - 6 | 962 | 77 |
GenBank mRNA | AJ582073.1 | - | 5 - 6 | 876 | 77 |