Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptr-1
Entrez Gene ID
191746
Gene Name
SSD domain-containing protein
Gene Aliases
CELE_C24B5.3
Chromosome Location
Chr.V: 9177949 - 9181719 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072979.4 NP_505380.1 5 - 6 1370 111

View Full Product Details