Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-29A4
Entrez Gene ID
181926
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_B0331.1
Chromosome Location
Chr.V: 9654755 - 9658120 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073089.4 NP_505490.2 3 - 4 293 138

View Full Product Details