Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpt-1
Entrez Gene ID
179641
Gene Name
26S proteasome regulatory subunit 7
Gene Aliases
CELE_C52E4.4
Chromosome Location
Chr.V: 11980502 - 11982397 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073604.7 NP_506005.1 1 - 2 76 103

View Full Product Details