Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-63
Entrez Gene ID
184399
Gene Name
Serpentine receptor class delta-63
Gene Aliases
CELE_F13A7.2
Chromosome Location
Chr.V: 16392482 - 16393828 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074742.2 NP_507143.2 4 - 5 714 61

View Full Product Details