Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aman-1
Entrez Gene ID
180749
Gene Name
Alpha-mannosidase
Gene Aliases
CELE_F55D10.1
Chromosome Location
Chr.X: 4733095 - 4738592 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076410.4 NP_508811.1 15 - 16 2069 128

View Full Product Details