Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-21
Entrez Gene ID
181516
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_C33G3.3
Chromosome Location
Chr.X: 14064313 - 14067984 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077940.3 NP_510341.2 9 - 10 1184 146

View Full Product Details