Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SULT1D1
Entrez Gene ID
403637
Gene Name
sulfotransferase family, cytosolic, 1D, member 1
Gene Aliases
-
Chromosome Location
Chr.13: 59270054 - 59306285 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_532396.6 XP_532396.1 7 - 8 925 149
GenBank mRNA AY004331.1 - 6 - 7 775 149

View Full Product Details