Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASIP
Entrez Gene ID
492296
Gene Name
agouti signaling protein
Gene Aliases
agouti
Chromosome Location
Chr.24: 23338757 - 23393913 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007263.1 NP_001007264.1 2 - 3 298 73
XM_014106843.2 XP_013962318.1 2 - 3 338 73
XM_022408819.1 XP_022264527.1 2 - 3 279 73
XM_022408820.1 XP_022264528.1 2 - 3 743 73
GenBank mRNA AY714374.1 - 2 - 3 298 73

View Full Product Details