Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAIM
Entrez Gene ID
485687
Gene Name
Fas apoptotic inhibitory molecule
Gene Aliases
-
Chromosome Location
Chr.23: 34807093 - 34822577 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005634497.3 XP_005634554.1 2 - 3 144 93
XM_005634499.3 XP_005634556.1 2 - 3 259 93
XM_022408685.1 XP_022264393.1 2 - 3 260 93
XM_542807.6 XP_542807.3 2 - 3 255 93

View Full Product Details