Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H12C
Entrez Gene ID
489416
Gene Name
zinc finger CCCH-type containing 12C
Gene Aliases
-
Chromosome Location
Chr.5: 22666321 - 22756089 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005619810.3 XP_005619867.1 4 - 5 3473 94
XM_005619811.3 XP_005619868.1 6 - 7 1543 94
XM_014113438.2 XP_013968913.1 6 - 7 1606 94
XM_022418342.1 XP_022274050.1 6 - 7 2090 94
XM_022418344.1 XP_022274052.1 7 - 8 1711 94
XM_022418345.1 XP_022274053.1 5 - 6 1438 94

View Full Product Details