Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDF2L1
Entrez Gene ID
608633
Gene Name
stromal cell derived factor 2 like 1
Gene Aliases
-
Chromosome Location
Chr.26: 30918788 - 30920873 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_845717.5 XP_850810.2 2 - 3 434 71

View Full Product Details