Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM97
Entrez Gene ID
610900
Gene Name
transmembrane protein 97
Gene Aliases
-
Chromosome Location
Chr.9: 42572855 - 42581012 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_848469.5 XP_853562.2 1 - 2 206 66

View Full Product Details