Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SC5D
Entrez Gene ID
610411
Gene Name
sterol-C5-desaturase
Gene Aliases
SC5DL
Chromosome Location
Chr.5: 12760591 - 12774484 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847911.4 XP_853004.1 - 83 62

View Full Product Details