Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZW10
Entrez Gene ID
479433
Gene Name
zw10 kinetochore protein
Gene Aliases
-
Chromosome Location
Chr.5: 19496531 - 19527205 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_536569.7 XP_536569.4 - 1595 69

View Full Product Details