Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACC1
Entrez Gene ID
475581
Gene Name
transforming acidic coiled-coil containing protein 1
Gene Aliases
-
Chromosome Location
Chr.16: 26677717 - 26797236 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005629845.3 XP_005629902.1 - 2421 137
XM_005629846.3 XP_005629903.1 - 2384 137
XM_014120038.2 XP_013975513.1 - 2300 137
XM_014120039.2 XP_013975514.1 - 2119 137
XM_022404033.1 XP_022259741.1 - 2509 137
XM_022404035.1 XP_022259743.1 - 2082 137
XM_022404036.1 XP_022259744.1 - 1995 137
XM_022404037.1 XP_022259745.1 - 2146 137
XM_022404038.1 XP_022259746.1 - 2002 137
XM_022404039.1 XP_022259747.1 - 1161 137
XM_022404040.1 XP_022259748.1 - 783 137
XM_843572.5 XP_848665.1 - 2472 137
XM_851541.5 XP_856634.1 - 1250 137

View Full Product Details