Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
LTBR
Entrez Gene ID
486728
Gene Name
lymphotoxin beta receptor
Gene Aliases
-
Chromosome Location
Chr.27: 38598725 - 38605829 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005637256.3 XP_005637313.1 3 - 3 539 137

View Full Product Details