Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPR50
Entrez Gene ID
492213
Gene Name
G protein-coupled receptor 50
Gene Aliases
-
Chromosome Location
Chr.X: 119266369 - 119271586 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_549333.4 XP_549333.3 2 - 2 857 66

View Full Product Details