Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIPK2B
Entrez Gene ID
612457
Gene Name
divergent protein kinase domain 2B
Gene Aliases
CXHXorf36
Chromosome Location
Chr.X: 38996003 - 39039589 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_850186.5 XP_855279.2 1 - 1 440 124

View Full Product Details