Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BNIPL
Entrez Gene ID
483190
Gene Name
BCL2 interacting protein like
Gene Aliases
-
Chromosome Location
Chr.17: 60126341 - 60135209 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_540308.6 XP_540308.3 4 - 4 801 94

View Full Product Details