Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abhd1
Entrez Gene ID
100763847
Gene Name
abhydrolase domain containing 1
Gene Aliases
I79_002569
Chromosome Location
Chr.Un|NW_003613640: 2142197 - 2149873 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497041.2 XP_003497089.1 1 - 2 2536 84

View Full Product Details