Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
En1
Entrez Gene ID
103163841
Gene Name
engrailed homeobox 1
Gene Aliases
I79_005518
Chromosome Location
Chr.Un|NW_003613737: 1194640 - 1204878 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640208.1 XP_007638398.1 3 - 4 492 95

View Full Product Details