Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fcamr
Entrez Gene ID
100773580
Gene Name
Fc receptor, IgA, IgM, high affinity
Gene Aliases
I79_003371
Chromosome Location
Chr.Un|NW_003613661: 2869542 - 2880803 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007616739.1 XP_007614929.1 1 - 2 163 59

View Full Product Details