Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a41
Entrez Gene ID
100762518
Gene Name
solute carrier family 25, member 41
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613861: 1217605 - 1224553 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642437.1 XP_007640627.1 1 - 2 286 73

View Full Product Details