Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cplx2
Entrez Gene ID
100768790
Gene Name
complexin 2
Gene Aliases
I79_003926
Chromosome Location
Chr.Un|NW_003613681: 1835746 - 1837485 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007623723.1 XP_007621913.1 4 - 5 465 93

View Full Product Details