Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbx6
Entrez Gene ID
100770940
Gene Name
T-box 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614830: 93428 - 97857 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510100.2 XP_003510148.1 5 - 6 1197 72

View Full Product Details