Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mfap2
Entrez Gene ID
100770393
Gene Name
microfibrillar-associated protein 2
Gene Aliases
I79_022282
Chromosome Location
Chr.Un|NW_003615865: 30287 - 32884 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513336.1 XP_003513384.1 7 - 8 462 69

View Full Product Details